Galβ(1-4)Glc-β-MP ≥96.0% (by HPLC)
Supplier: TCI America
Synonyms:
LacMP
, 4-Methoxyphenyl 4-O-(β-D-Galactopyranosyl)-β-D-glucopyranoside
M1805-1G
TCM1805-1GEA
211.93
USD
TCM1805-1G
Galβ(1-4)Glc-β-MP ≥96.0% (by HPLC)
Gal beta(1-4)Glc-beta-MP
CAS Number: 150412-80-9
Molecular Formula: C19H28O12
Molecular Weight: 448.42
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: -21 deg (C=0.5, DMF)
Storage Temperature:
Product Categories: Synthetic Organic Chemistry, Glycosylation, Bioscience, Disaccharides, Glycosides, p-Methoxyphenyl Glycosides
Other Product Names / Synonyms: 4-Methoxyphenyl 4-O-(beta-D-Galactopyranosyl)-beta-D-glucopyranoside, LacMP
Formula:
C₁₉H₂₈O₁₂ MW: 448.42 g/mol |
CAS Number:
150412-80-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...