Methyl-2-ethoxybenzoate ≥99.0%
Supplier: TCI America
Synonyms:
Methyl 2-ethoxybenzoate
M1785-25G
TCM1785-25GEA
116
USD
TCM1785-25G
Methyl-2-ethoxybenzoate ≥99.0%
Methyl 2-ethoxybenzoate
CAS Number: 3686-55-3
MDL Number: MFCD00017261
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 245
Specific Gravity (20/20): 1.11
Other Product Names / Synonyms: 2-Ethoxybenzoic Acid Methyl Ester
Formula:
C₁₀H₁₂O₃ MW: 180.2005 g/mol Boiling Pt: 245 °C Density: 1.11 (20 °C) |
MDL Number:
MFCD00017261 CAS Number: 3686-55-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...