3-(3-Methoxyphenoxy)propionic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
3-(3-Methoxyphenoxy)propionic Acid
M1692-5G
TCM1692-5GEA
192
USD
TCM1692-5G
3-(3-Methoxyphenoxy)propionic acid ≥97.0% (by GC, titration analysis)
3-(3-Methoxyphenoxy)propionic acid
CAS Number: 49855-03-0
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 83
Formula:
C₁₀H₁₂O₄ MW: 196.1999 g/mol Melting Pt: 83 °C |
MDL Number:
MFCD00045943 CAS Number: 49855-03-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...