3'-(Methylthio)acetanilide

Supplier: TCI America

Synonyms: 3-Acetamidothioanisole

M1671-25G
TCM1671-25GEA 146.11 USD
TCM1671-25G
3'-(Methylthio)acetanilide
3'-(Methylthio)acetanilide

CAS Number: 2524-78-9
Molecular Formula: C9H11NOS
Molecular Weight: 181.25
Form: Crystal
Color: Pale Yellowish Gray


Other Product Names / Synonyms: 3-Acetamidothioanisole

https://us.vwr.com/stibo/web/std.lang.all/59/60/28975960.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR