2-Methyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1,3-Diamino-2-methylpropane
M1657-5G
M1657-1G
TCM1657-5GEA
312.53
USD
TCM1657-5G
TCM1657-1G
2-Methyl-1,3-propanediamine ≥98.0% (by GC, titration analysis)
2-Methyl-1,3-propanediamine
CAS Number: 2400-78-4
Molecular Formula: C4H12N2
Molecular Weight: 88.15
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 0.88
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3-Diamino-2-methylpropane
Formula:
C₄H₁₂N₂ MW: 88.15 g/mol Density: 0.88 (20 °C) |
CAS Number:
2400-78-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...