2-(4-Methoxyphenyl)benzothiazole ≥98.0%

Supplier: TCI America

Synonyms: 1-(2-Benzothiazolyl)-4-methoxybenzene4-(2-Benzothiazolyl)anisole

M1654-5G
TCM1654-5GEA 131 USD
TCM1654-5G
2-(4-Methoxyphenyl)benzothiazole ≥98.0%
2-(4-Methoxyphenyl)benzothiazole

CAS Number: 6265-92-5
Molecular Formula: C14H11NOS
Molecular Weight: 241.31
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 122


Other Product Names / Synonyms: 4-(2-Benzothiazolyl)anisole, 1-(2-Benzothiazolyl)-4-methoxybenzene

Formula: C₁₄H₁₁NOS
MW: 241.31 g/mol
Melting Pt: 122 °C
MDL Number: MFCD01099069
CAS Number: 6265-92-5
https://us.vwr.com/stibo/web/std.lang.all/59/29/28975929.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR