Mono-6-O-(p-toluenesulfonyl)-ɑ-cyclodextrin ≥85.0% (by HPLC)
Supplier: TCI America
M1644-1G
M1644-200MG
TCM1644-200MGEA
441
USD
TCM1644-200MG
TCM1644-1G
Mono-6-O-(p-toluenesulfonyl)-ɑ-cyclodextrin ≥85.0% (by HPLC)
Mono-6-O-(p-toluenesulfonyl)-alpha-cyclodextrin
CAS Number: 32860-56-3
Molecular Formula: C43H66O32S
Molecular Weight: 1127.03
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: 122 deg (C=4, DMSO)
Storage Temperature:
Formula:
C₄₃H₆₆O₃₂S MW: 1127.03 g/mol |
CAS Number:
32860-56-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...