4'-(Methoxycarbonyl)benzo-15-crown-5 (4'-(Methoxycarbonyl)benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥96.0%
Synonyms:
2,3-(4-Methoxycarbonylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
, 4'-Methoxycarbonylbenzo-15-crown 5-Ether
M1489-1G
TCM1489-1GEA
618
USD
TCM1489-1G
4'-(Methoxycarbonyl)benzo-15-crown-5 (4'-(Methoxycarbonyl)benzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥96.0%
4'-(Methoxycarbonyl)benzo-15-crown-5 (4'-(Methoxycarbonyl)benzo-1,4,7,10,13-pentaoxacyclopentadecane)
CAS Number: 56683-56-8
MDL Number: MFCD06797129
Molecular Formula: C16H22O7
Molecular Weight: 326.35
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 82
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-(4-Methoxycarbonylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
Formula:
C₁₆H₂₂O₇ Melting Pt: 82 °C |
MDL Number:
MFCD06797129 CAS Number: 56683-56-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...