(1S,2S)-2-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

A1689-100MG
TCA1689-100MGEA 127.5 USD
TCA1689-100MG
(1S,2S)-2-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
(1S,2S)-2-Aminocyclohexanecarboxylic acid

CAS Number: 24716-93-6
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Specific rotation [a]20/D: 76.5 deg (C=1, H2O)

Formula: C₇H₁₃NO₂
MDL Number: MFCD01318270
CAS Number: 24716-93-6
https://us.vwr.com/stibo/web/std.lang.all/98/74/28929874.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR