1-(Methoxymethoxy)naphthalene ≥97.0%

Supplier: TCI America

M1454-5G
TCM1454-5GEA 105.53 USD
TCM1454-5G
1-(Methoxymethoxy)naphthalene ≥97.0%
1-(Methoxymethoxy)naphthalene

CAS Number: 7382-37-8
MDL Number: MFCD06797119
Molecular Formula: C12H12O2
Molecular Weight: 188.23
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 296
Specific Gravity (20/20): 1.12

Formula: C₁₂H₁₂O₂
Boiling Pt: 296 °C
Density: 1.12 (20 °C)
MDL Number: MFCD06797119
CAS Number: 7382-37-8
https://us.vwr.com/stibo/web/std.lang.all/56/46/28975646.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR