2-Methoxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1417-25G M1417-5G
TCM1417-25GEA 357.41 USD
TCM1417-25G TCM1417-5G
2-Methoxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)
2-Methoxy-1-naphthoic acid

CAS Number: 947-62-6
MDL Number: MFCD00021532
Molecular Formula: C12H10O3
Molecular Weight: 202.21
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 178

https://us.vwr.com/stibo/web/std.lang.all/55/63/28975563.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR