1,1'-Methylenedi-2-naphthol ≥97.0%
Supplier: TCI America
Synonyms:
Bis(2-hydroxy-1-naphthyl)methane
, 1,1'-Methylenebis(2-naphthol)
M1410-25G
TCM1410-25GEA
224.26
USD
TCM1410-25G
1,1'-Methylenedi-2-naphthol ≥97.0%
1,1'-Methylenedi-2-naphthol
CAS Number: 1096-84-0
MDL Number: MFCD00046470
Molecular Formula: C21H16O2
Molecular Weight: 300.36
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Bis(2-hydroxy-1-naphthyl)methane, 1,1'-Methylenebis(2-naphthol)
Formula:
C₂₁H₁₆O₂ MW: 300.36 g/mol |
MDL Number:
MFCD00046470 CAS Number: 1096-84-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...