γ-Methylene-γ-butyrolactone ≥98.0%

Supplier: TCI America

Synonyms: 5-Methylenedihydro-2(5H)-furanone

M1385-1G
TCM1385-1GEA 284 USD
TCM1385-1G
γ-Methylene-γ-butyrolactone ≥98.0%
gamma-Methylene-gamma-butyrolactone

CAS Number: 10008-73-8
MDL Number: MFCD00042762
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 77
Specific Gravity (20/20): 1.11


Other Product Names / Synonyms: 5-Methylenedihydro-2(5H)-furanone

Formula: C₅H₆O₂
MW: 98.10 g/mol
Density: 1.11 (20 °C)
Flash Pt: 77 °C
MDL Number: MFCD00042762
CAS Number: 10008-73-8
https://us.vwr.com/stibo/web/std.lang.all/55/03/28975503.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR