Mono-6-O-(p-toluenesulfonyl)-β-cyclodextrin ≥85.0% (by HPLC)

Synonyms: Mono-(6-p-toluenesulphonyl)-β-cyclodextrin

M1381-200MG
TCM1381-200MGEA 365.65 USD
TCM1381-200MG
Mono-6-O-(p-toluenesulfonyl)-β-cyclodextrin ≥85.0% (by HPLC)
Mono-6-O-(p-toluenesulfonyl)-beta-cyclodextrin

CAS Number: 67217-55-4
Molecular Weight: 1289.17
Purity/Analysis Method: 85.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 139 deg (C=4, DMSO)
Storage Temperature: 0-10°C

Formula: C₄₉H₇₆O₃₇S
MW: 1289.1706 g/mol
MDL Number: MFCD05864975
CAS Number: 67217-55-4
https://us.vwr.com/stibo/web/std.lang.all/54/52/28975452.jpg

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR