1,3,5-Adamantanetriol ≥97.0%
Supplier: TCI America
Synonyms:
Adamantane-1,3,5-triol
, 1,3,5-Trihydroxyadamantane
A1682-250MG
A1682-5G
TCA1682-250MGEA
41.6
USD
TCA1682-250MG
TCA1682-5G
1,3,5-Adamantanetriol ≥97.0%
1,3,5-Adamantanetriol
CAS Number: 99181-50-7
Molecular Formula: C10H16O3
Molecular Weight: 184.24
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,3,5-Trihydroxyadamantane
Formula:
C₁₀H₁₆O₃ MW: 184.24 g/mol |
MDL Number:
MFCD08276262 CAS Number: 99181-50-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...