(S)-(+)-2-Methoxy-2-(1-naphthyl)propionic acid ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: (S)-(+)-M alpha NP Acid(S)-(+)-alpha-Methoxy-alpha-methyl-1-naphthaleneacetic Acid

M1367-100MG
TCM1367-100MGEA 312 USD
TCM1367-100MG
(S)-(+)-2-Methoxy-2-(1-naphthyl)propionic acid ≥99.0% (by HPLC)
(S)-(+)-2-Methoxy-2-(1-naphthyl)propionic acid

CAS Number: 102691-93-0
MDL Number: MFCD03093634
Molecular Formula: C14H14O3
Molecular Weight: 230.26
Purity/Analysis Method: 99.0% (HPLC)
Form: Crystal
Melting point (°C): 110
Specific rotation [a]20/D: 100 deg (C=1, MeOH)


Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (S)-(+)-alpha-Methoxy-alpha-methyl-1-naphthaleneacetic Acid, (S)-(+)-M alpha NP Acid

Formula: C₁₄H₁₄O₃
MW: 230.26 g/mol
Melting Pt: 110 °C
MDL Number: MFCD03093634
CAS Number: 102691-93-0
https://us.vwr.com/stibo/web/std.lang.all/54/23/28975423.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR