(R)-(-)-2-Methoxy-2-(1-naphthyl)propionic acid ≥99.0% (by HPLC)

Supplier: TCI America

Synonyms: (R)-(-)-alpha-Methoxy-alpha-methyl-1-naphthaleneacetic Acid(R)-(?)-M alpha NP Acid

M1366-100MG
TCM1366-100MGEA 312 USD
TCM1366-100MG
(R)-(-)-2-Methoxy-2-(1-naphthyl)propionic acid ≥99.0% (by HPLC)
(R)-(-)-2-Methoxy-2-(1-naphthyl)propionic acid

CAS Number: 63628-26-2
MDL Number: MFCD03093633
Molecular Formula: C14H14O3
Molecular Weight: 230.26
Purity/Analysis Method: 99.0% (HPLC)
Form: Crystal
Melting point (°C): 108
Specific rotation [a]20/D: -100 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Analytical Science, e.e. Determination (HPLC Labeling Reagents), e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R)-(-)-alpha-Methoxy-alpha-methyl-1-naphthaleneacetic Acid, (R)-(?)-M alpha NP Acid

Formula: C₁₄H₁₄O₃
MW: 230.26 g/mol
Melting Pt: 108 °C
MDL Number: MFCD03093633
CAS Number: 63628-26-2
https://us.vwr.com/stibo/web/std.lang.all/54/18/28975418.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR