Montanic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Octacosanoic acidMontanic acid

M1342-100MG
TCM1342-100MGEA 54.81 USD
TCM1342-100MG
Montanic acid ≥98.0% (by titrimetric analysis)
Montanic acid

CAS Number: 506-48-9
MDL Number: MFCD00002812
Molecular Formula: C28H56O2
Molecular Weight: 424.75
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 89


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Montanic Acid

Formula: C₂₈H₅₆O₂
MW: 424.75 g/mol
Boiling Pt: 250 °C (0.01 mmHg)
Melting Pt: 89 °C
Density: 0.8191 g/cm³ (100 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002812
CAS Number: 506-48-9
EINECS: 208-041-8
https://us.vwr.com/stibo/web/std.lang.all/53/18/28975318.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR