5-(4-Methoxycarbonylphenyl)-10,15,20-triphenylporphyrin ≥90.0% (by HPLC)
Supplier: TCI America
Synonyms:
5-(4-Carbomethoxyphenyl)-10,15,20-triphenylporphyrin
, 5-(4-Methoxycarbonylphenyl)-10,15,20-triphenyl-21H,23H-porphine
CAS Number: 119730-06-2
MDL Number: MFCD02093480
Molecular Formula: C46H32N4O2
Molecular Weight: 672.79
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins, Analytical Science, Absolute Configuration Determination (Exciton Chirality CD Method)
Other Product Names / Synonyms: 5-(4-Carbomethoxyphenyl)-10,15,20-triphenylporphyrin, 5-(4-Methoxycarbonylphenyl)-10,15,20-triphenyl-21H,23H-porphine
Formula:
C₄₆H₃₂N₄O₂ MW: 672.79 g/mol |
MDL Number:
MFCD02093480 CAS Number: 119730-06-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...