3-Methylpent-1-en-4-yn-3-ol ≥98.0%

Supplier: TCI America

M1312-5ML M1312-25ML
TCM1312-005MLEA 119 USD
TCM1312-005ML TCM1312-025ML
3-Methylpent-1-en-4-yn-3-ol ≥98.0%
3-Methylpent-1-en-4-yn-3-ol

CAS Number: 3230-69-1
MDL Number: MFCD01709006
Molecular Formula: C6H8O
Molecular Weight: 96.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 32
Specific Gravity (20/20): 0.89

Formula: C₆H₈O
Boiling Pt: 121 °C
Density: 0.89 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD01709006
CAS Number: 3230-69-1
EINECS: 221-770-6
UN: 1987
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/51/99/28975199.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR