2-Isopropoxyethyl methanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Methanesulfonic acid 2-Isopropoxyethyl EsterMethanesulphonic acid 2-Isopropoxyethyl EsterMEthanesulfonic acid 2-isopropoxyethyl esterMEthanesulphonic acid 2-isopropoxyethyl ester

M1297-25G
TCM1297-025GEA 255.67 USD
TCM1297-025G
2-Isopropoxyethyl methanesulfonate ≥98.0%
2-Isopropoxyethyl methanesulfonate

CAS Number: 235097-76-4
MDL Number: MFCD02093478
Molecular Formula: C6H14O4S
Molecular Weight: 182.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.14


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Methanesulfonic Acid 2-Isopropoxyethyl Ester

Formula: C₆H₁₄O₄S
MW: 182.23 g/mol
Density: 1.14 (20 °C)
MDL Number: MFCD02093478
CAS Number: 235097-76-4
https://us.vwr.com/stibo/web/std.lang.all/51/72/28975172.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR