β-Methylcholine chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Hydroxypropyltrimethylammonium Chloride
M1293-25G
TCM1293-025GEA
147.11
USD
TCM1293-025G
β-Methylcholine chloride ≥98.0% (by titrimetric analysis)
beta-Methylcholine chloride
CAS Number: 2382-43-6
MDL Number: MFCD01669408
Molecular Formula: C6H16ClNO
Molecular Weight: 153.65
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: 2-Hydroxypropyltrimethylammonium Chloride
Formula:
C₆H₁₆ClNO MW: 153.65 g/mol |
MDL Number:
MFCD01669408 CAS Number: 2382-43-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...