3-Methyl-1-hexanol ≥97.0%

Supplier: TCI America

M1246-5G M1246-1G
TCM1246-5GEA 377 USD
TCM1246-5G TCM1246-001G
3-Methyl-1-hexanol ≥97.0%
3-Methyl-1-hexanol

CAS Number: 13231-81-7
MDL Number: MFCD01321178
Molecular Formula: C7H16O
Molecular Weight: 116.20
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Specific Gravity (20/20): 0.83

Formula: C₇H₁₆O
Boiling Pt: 96 °C
Density: 0.83 (20 °C)
MDL Number: MFCD01321178
CAS Number: 13231-81-7
https://us.vwr.com/stibo/web/std.lang.all/50/43/28975043.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR