Dimethyl (S)-(-)-methylsuccinate ≥98.0%

Supplier: TCI America

Synonyms: Dimethyl (S)-(-)-Pyrotartarate(S)-(-)-Methylsuccinic Acid Dimethyl Ester(S)-(-)-Pyrotartaric Acid Dimethyl Ester

M1232-5G
TCM1232-005GEA 205.95 USD
TCM1232-005G
Dimethyl (S)-(-)-methylsuccinate ≥98.0%
Dimethyl (S)-(-)-methylsuccinate

CAS Number: 63163-08-6
MDL Number: MFCD00134572
Molecular Formula: C7H12O4
Molecular Weight: 160.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 83
Specific Gravity (20/20): 1.08
Specific rotation [a]20/D: -6 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: Dimethyl (S)-(-)-Pyrotartarate, (S)-(-)-Methylsuccinic Acid Dimethyl Ester, (S)-(-)-Pyrotartaric Acid Dimethyl Ester

Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 196 °C
Density: 1.08 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00134572
CAS Number: 63163-08-6
https://us.vwr.com/stibo/web/std.lang.all/50/18/28975018.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR