5-Methyl-2-heptanol

Supplier: TCI America

M1222-5ML
TCM1222-005MLEA 182 USD
TCM1222-005ML
5-Methyl-2-heptanol
5-Methyl-2-heptanol

CAS Number: 54630-50-1
MDL Number: MFCD00060896
Molecular Formula: C8H18O
Molecular Weight: 130.23
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 172
Flash Point (°C): 38
Specific Gravity (20/20): 0.83

Formula: C₈H₁₈O
Boiling Pt: 172 °C
Density: 0,83 (20 °C)
Flash Pt: 38 °C
MDL Number: MFCD00060896
CAS Number: 54630-50-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR