(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(6-Methoxynaphthalen-2-yl)propanoic acid(+/-)-Naproxen(+/-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid

M1220-25G
TCM1220-025GEA 307.55 USD
TCM1220-025G
(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid ≥98.0% (by GC, titration analysis)
(+/-)-2-(6-Methoxy-2-naphthyl)propionic acid

CAS Number: 23981-80-8
MDL Number: MFCD00439456
Molecular Formula: C14H14O3
Molecular Weight: 230.26
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 157


Product Categories: Bioscience, Immune System, Inflammation, Allergy Research Reagents, Anti-inflammatory Ingredients
Other Product Names / Synonyms: (+/-)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid, (+/-)-Naproxen

Formula: C₁₄H₁₄O₃
MW: 230.26 g/mol
Melting Pt: 157 °C
MDL Number: MFCD00439456
CAS Number: 23981-80-8
https://us.vwr.com/stibo/web/std.lang.all/50/00/28975000.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR