Mono-6-O-mesitylenesulphonyl-γ-cyclodextrin ≥90.0% (by HPLC)

Supplier: TCI America

M1212-1G
TCM1212-001GEA 587 USD
TCM1212-001G
Mono-6-O-mesitylenesulphonyl-γ-cyclodextrin ≥90.0% (by HPLC)
Mono-6-O-mesitylenesulfonyl-gamma-cyclodextrin

CAS Number: 174010-62-9
MDL Number: MFCD00671559
Molecular Formula: C57H90O42S
Molecular Weight: 1479.37
Purity/Analysis Method: 90.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 139 deg (C=1, H2O)

Formula: C₅₇H₉₀O₄₂S
MW: 1479.37 g/mol
MDL Number: MFCD00671559
CAS Number: 174010-62-9
https://us.vwr.com/stibo/web/std.lang.all/49/88/28974988.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR