N-(5-Aminopentyl)-2-nitrobenzenesulfonamide hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N-(o-Ns)-1,5-diaminopentane Hydrochloride
, 1-Amino-5-(2-nitrobenzenesulfonamido)pentane Hydrochloride
, 1-Amino-5-(2-nitrobenzenesulphonamido)pentane Hydrochloride
, N-(2-Nitrobenzenesulphonyl)-1,5-pentanediamine Hydrochloride
, N-(2-Nitrobenzenesulfonyl)-1,5-pentanediamine Hydrochloride
CAS Number: 437718-20-2
MDL Number: MFCD06797044
Molecular Formula: C11H17N3O4S
Molecular Weight: 323.79
Purity/Analysis Method:
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: 1-Amino-5-(2-nitrobenzenesulfonamido)pentane Hydrochloride, N-(2-Nitrobenzenesulfonyl)-1,5-pentanediamine Hydrochloride, N-(o-Ns)-1,5-diaminopentane Hydrochloride
Formula:
C₁₁H₁₇N₃O₄S MW: 323.79 g/mol |
MDL Number:
MFCD06797044 CAS Number: 437718-20-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...