3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one ≥98.0%

Supplier: TCI America

M1188-1G
TCM1188-001GEA 77.11 USD
TCM1188-001G
3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one ≥98.0%
3-(Methoxycarbonyl)-2H-cyclohepta[b]furan-2-one

CAS Number: 50603-71-9
MDL Number: MFCD00671552
Molecular Formula: C11H8O4
Molecular Weight: 204.18
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 173

Formula: C₁₁H₈O₄
MW: 204.18 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00671552
CAS Number: 50603-71-9
https://us.vwr.com/stibo/web/std.lang.all/49/36/28974936.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR