cis-β-Methylstyrene ≥98.0% stabilized
Supplier: TCI America
Synonyms:
cis-1-Phenyl-1-propene
M1174-10ML
TCM1174-010MLEA
317.74
USD
TCM1174-010ML
cis-β-Methylstyrene ≥98.0% stabilized
cis-beta-Methylstyrene
(stabilized with TBC)
CAS Number: 766-90-5
MDL Number: MFCD00065137
Molecular Formula: C9H10
Molecular Weight: 118.18
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 174
Melting point (°C): -60
Flash Point (°C): 45
Specific Gravity (20/20): 0.91
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: cis-1-Phenyl-1-propene, cis-Propenylbenzene
Formula:
C₉H₁₀ MW: 118.18 g/mol Boiling Pt: 174 °C Melting Pt: –60 °C Density: 0.91 (20 °C) Flash Pt: 45 °C |
MDL Number:
MFCD00065137 CAS Number: 766-90-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...