(1S,2S)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic acid ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: N-[(1S,2S)-2-Carboxycyclohexyl]anthracene-2,3-dicarboximide

A1658-100MG
TCA1658-100MGEA 402 USD
TCA1658-100MG
(1S,2S)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic acid ≥97.0% (by HPLC)
(1S,2S)-2-(Anthracene-2,3-dicarboximido)cyclohexanecarboxylic acid

CAS Number: 446044-45-7
Molecular Formula: C23H19NO4
Molecular Weight: 373.41
Purity/Analysis Method: 97.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 42 deg (C=0.2, THF)


Product Categories: Analytical Science, e.e. Determination (HPLC Labeling Reagents), e.e. / Absolute Configuration Determination (NMR), Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: N-[(1S,2S)-2-Carboxycyclohexyl]anthracene-2,3-dicarboximide

Formula: C₂₃H₁₉NO₄
MW: 373.41 g/mol
CAS Number: 446044-45-7
https://us.vwr.com/stibo/web/std.lang.all/98/33/28929833.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR