(R)-(+)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde ≥98.0%

Supplier: TCI America

Synonyms: (R)-(+)-1-Formyl-2-(methoxymethyl)pyrrolidine

M1170-1G
TCM1170-001GEA 62.9 USD
TCM1170-001G
(R)-(+)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde ≥98.0%
(R)-(+)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde

CAS Number: 121817-71-8
MDL Number: MFCD00191670
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.06
Specific rotation [a]20/D: 35 deg (C=2, Toluene)


Other Product Names / Synonyms: (R)-(+)-1-Formyl-2-(methoxymethyl)pyrrolidine

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Density: 1.06 (20 °C)
MDL Number: MFCD00191670
CAS Number: 121817-71-8
https://us.vwr.com/stibo/web/std.lang.all/48/96/28974896.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR