2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile ≥98.0%

Supplier: TCI America

Synonyms: alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile

M1163-5G
TCM1163-005GEA 56.79 USD
TCM1163-005G
2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile ≥98.0%
2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile

CAS Number: 80866-87-1
MDL Number: MFCD00001844
Molecular Formula: C10H8F3NO
Molecular Weight: 215.18
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 67
Specific Gravity (20/20): 1.15


Other Product Names / Synonyms: alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile

Formula: C₁₀H₈F₃NO
MW: 215.18 g/mol
Boiling Pt: 82 °C
Density: 1.15 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00001844
CAS Number: 80866-87-1
https://us.vwr.com/stibo/web/std.lang.all/48/84/28974884.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR