(S)-(-)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde ≥96.0%

Synonyms: (S)-(-)-1-Formyl-2-(methoxymethyl)pyrrolidine

M1162-1G
TCM1162-001GEA 92.7 USD
TCM1162-001G
(S)-(-)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde ≥96.0%
(S)-(-)-2-(Methoxymethyl)-1-pyrrolidinecarboxaldehyde

CAS Number: 63126-45-4
MDL Number: MFCD00010279
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: (S)-(-)-1-Formyl-2-(methoxymethyl)pyrrolidine

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00010279
CAS Number: 63126-45-4
https://us.vwr.com/stibo/web/std.lang.all/48/82/28974882.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR