Diethyl-4-methoxyphenylphosphonate ≥97.0%

Supplier: TCI America

Synonyms: 4-Methoxyphenylphosphonic Acid Diethyl Ester

M1152-10G
TCM1152-010GEA 244.52 USD
TCM1152-010G
Diethyl-4-methoxyphenylphosphonate ≥97.0%
Diethyl-4-methoxyphenylphosphonate

CAS Number: 3762-33-2
MDL Number: MFCD00015670
Molecular Formula: C11H17O4P
Molecular Weight: 244.23
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid


Other Product Names / Synonyms: 4-Methoxyphenylphosphonic Acid Diethyl Ester

Formula: C₁₁H₁₇O₄P
MW: 244.23 g/mol
MDL Number: MFCD00015670
CAS Number: 3762-33-2
https://us.vwr.com/stibo/web/std.lang.all/48/64/28974864.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR