TCIAM1147

M1147-10ML
TCM1147-010MLEA 95.74 USD
TCM1147-010ML
TCIAM1147
CAS Number: 28897-58-7
MDL Number: MFCD00015855
Molecular Formula: C7H12O2
Molecular Weight: 128.17
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 89
Flash Point (°C): 86
Specific Gravity (20/20): 0.97
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR