Ethyl-2-methyl-3-furancarboxylate ≥96.0%

Supplier: TCI America

Synonyms: 2-Methyl-3-furoic Acid Ethyl EsterEthyl 2-Methyl-3-furoate2-Methyl-3-furancarboxylic Acid Ethyl Ester

M1140-10G
TCM1140-010GEA 244 USD
TCM1140-010G
Ethyl-2-methyl-3-furancarboxylate ≥96.0%
Ethyl-2-methyl-3-furancarboxylate

CAS Number: 28921-35-9
MDL Number: MFCD00134327
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Flash Point (°C): 71
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 2-Methyl-3-furancarboxylic Acid Ethyl Ester, Ethyl 2-Methyl-3-furoate, 2-Methyl-3-furoic Acid Ethyl Ester

Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 87 °C
Density: 1.08 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00134327
CAS Number: 28921-35-9
https://us.vwr.com/stibo/web/std.lang.all/48/45/28974845.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR