7-Methoxy-3-phenylsulfonyl-1(3H)-isobenzofuranone ≥98.0% (by HPLC)

Supplier: TCI America

M1130-1G
TCM1130-001GEA 238 USD
TCM1130-001G
7-Methoxy-3-phenylsulfonyl-1(3H)-isobenzofuranone ≥98.0% (by HPLC)
7-Methoxy-3-phenylsulfonyl-1(3H)-isobenzofuranone

CAS Number: 65131-09-1
MDL Number: MFCD00191458
Molecular Formula: C15H12O5S
Molecular Weight: 304.32
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 188

Formula: C₁₅H₁₂O₅S
MDL Number: MFCD00191458
CAS Number: 65131-09-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR