(3S)-(-)-3-(Methylamino)pyrrolidine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

M1108-5G M1108-1G
TCM1108-5GEA 496 USD
TCM1108-5G TCM1108-001G
(3S)-(-)-3-(Methylamino)pyrrolidine ≥97.0% (by GC, titration analysis)
(3S)-(-)-3-(Methylamino)pyrrolidine

CAS Number: 139015-32-0
MDL Number: MFCD00191462
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 164
Specific Gravity (20/20): 0.94
Specific rotation [a]20/D: -20 deg (C=10, EtOH)

Formula: C₅H₁₂N₂
Boiling Pt: 164 °C
Density: 0.94 (20 °C)
MDL Number: MFCD00191462
CAS Number: 139015-32-0
https://us.vwr.com/stibo/web/std.lang.all/47/94/28974794.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR