3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M1106-5G M1106-25G
TCM1106-25GEA 930.18 USD
TCM1106-25G TCM1106-005G
3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
3-(Methylamino)pyrrolidine

CAS Number: 83030-08-4
MDL Number: MFCD00191460
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 165
Specific Gravity (20/20): 0.94

Formula: C₅H₁₂N₂
MW: 100.17 g/mol
Boiling Pt: 165 °C
Density: 0.94 (20 °C)
MDL Number: MFCD00191460
CAS Number: 83030-08-4
https://us.vwr.com/stibo/web/std.lang.all/47/90/28974790.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR