TCIAM1075

M1075-1G
TCM1075-001GEA 151.39 USD
TCM1075-001G
TCIAM1075
CAS Number: 2716-53-2
MDL Number: MFCD00069635
Molecular Formula: C21H40O4
Molecular Weight: 356.55
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 58

Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Glycerol alpha-Monoelaidate, 1-Mono-trans-9-octadecenoyl Glycerol
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR