TCIAA1630

A1630-1G
TCA1630-1GEA 341.71 USD
TCA1630-1G
TCIAA1630
CAS Number: 211512-13-9
MDL Number: MFCD04117909
Molecular Formula: C10H15N3O4S
Molecular Weight: 273.31
Purity/Analysis Method: 95.0% (HPLC,T)
Form: Crystal

Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Bioscience, Chemical Building Blocks for Polyamines
Other Product Names / Synonyms: 1-Amino-4-(2-nitrobenzenesulfonamido)butane, N-(2-Nitrobenzenesulfonyl)-1,4-butanediamine, N-(o-Ns)-1,4-diaminobutane
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR