3-Methyl-3-heptene (cis- and trans- mixture) ≥97.0%

Supplier: TCI America

M0991-1ML M0991-5ML
TCM0991-001MLEA 182 USD
TCM0991-001ML TCM0991-005ML
3-Methyl-3-heptene (cis- and trans- mixture) ≥97.0%
3-Methyl-3-heptene (cis- and trans- mixture)

CAS Number: 7300-03-0
MDL Number: MFCD00048796
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Specific Gravity (20/20): 0.73

Formula: C₈H₁₆
MW: 112,22 g/mol
Boiling Pt: 121 °C
Density: 0,73 (20 °C)
MDL Number: MFCD00048796
CAS Number: 7300-03-0

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR