N-(2-Methoxyethyl)isopropylamine ≥98.0%
Supplier: TCI America
M0987-25ML
M0987-5ML
TCM0987-5MLEA
189.81
USD
TCM0987-5ML
TCM0987-25ML
N-(2-Methoxyethyl)isopropylamine ≥98.0%
N-(2-Methoxyethyl)isopropylamine
CAS Number: 104678-18-4
MDL Number: MFCD00144828
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 125
Specific Gravity (20/20): 0.82
Formula:
C₆H₁₅NO Boiling Pt: 125 °C Density: 0.82 (20 °C) |
MDL Number:
MFCD00144828 CAS Number: 104678-18-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...