N-(2-Methoxyethyl)propylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0980-25G M0980-5G
TCM0980-025GEA 329 USD
TCM0980-025G TCM0980-005G
N-(2-Methoxyethyl)propylamine ≥98.0% (by GC, titration analysis)
N-(2-Methoxyethyl)propylamine

CAS Number: 43175-57-1
MDL Number: MFCD00144830
Molecular Formula: C6H15NO
Molecular Weight: 117.19
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 138
Specific Gravity (20/20): 0.83

Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 138 °C
Density: 0.83 (20 °C)
MDL Number: MFCD00144830
CAS Number: 43175-57-1
https://us.vwr.com/stibo/web/std.lang.all/45/64/28974564.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR