TCIAM0973

M0973-10G
TCM0973-010GEA 164.75 USD
TCM0973-010G
TCIAM0973
CAS Number: 143085-87-4
MDL Number: MFCD00144820
Molecular Formula: C15H23NO
Molecular Weight: 233.36
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 52

Other Product Names / Synonyms: 3-Isovaleramido-1-phenylbutane
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR