(S)-(+)-3-Methyl-1-pentanol ≥98.0%
Supplier: TCI America
M0963-5ML
M0963-1ML
TCM0963-005MLEA
518.51
USD
TCM0963-005ML
TCM0963-001ML
(S)-(+)-3-Methyl-1-pentanol ≥98.0%
(S)-(+)-3-Methyl-1-pentanol
CAS Number: 42072-39-9
MDL Number: MFCD00135167
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 58
Specific Gravity (20/20): 0.83
Specific rotation [a]20/D: 8.7 deg (neat)
Formula:
C₆H₁₄O Boiling Pt: 152 °C Density: 0,83 (20 °C) Flash Pt: 58 °C |
MDL Number:
MFCD00135167 CAS Number: 42072-39-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...