Methyl-3-methoxypropionate ≥99.0%
Supplier: TCI America
Synonyms:
Methyl 3-methoxypropionate
M0962-500ML
M0962-25ML
TCM0962-500MLEA
165
USD
TCM0962-500ML
TCM0962-025ML
Methyl-3-methoxypropionate ≥99.0%
Methyl 3-methoxypropionate
CAS Number: 3852-09-3
MDL Number: MFCD00059085
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 47
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: 3-Methoxypropionic Acid Methyl Ester
Formula:
C₅H₁₀O₃ MW: 118.1311 g/mol Boiling Pt: 144 °C Density: 1.01 (20 °C) Flash Pt: 47 °C |
UN:
3272 ADR: 3,III |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...