D-Mannono-1,4-lactone ≥97.0%

Supplier: TCI America

Synonyms: (3S,4R,5R)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxydihydrofuran-2(3H)-one

M0958-1G M0958-5G
TCM0958-1GEA 35.68 USD
TCM0958-1G TCM0958-005G
D-Mannono-1,4-lactone ≥97.0%
D-Mannono-1,4-lactone

CAS Number: 26301-79-1
MDL Number: MFCD00065020
Molecular Formula: C6H10O6
Molecular Weight: 178.14
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 153
Specific rotation [a]20/D: 52 deg (C=2, H2O)

Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00065020
CAS Number: 26301-79-1
https://us.vwr.com/stibo/web/std.lang.all/45/29/28974529.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR