N-Methoxy-N-methylacetamide ≥98.0%
Supplier: TCI America
M0919-10G
M0919-25G
TCM0919-010GEA
46.67
USD
TCM0919-010G
TCM0919-25G
N-Methoxy-N-methylacetamide ≥98.0%
N-Methoxy-N-methylacetamide
CAS Number: 78191-00-1
MDL Number: MFCD00060098
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): 49
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: N-Acetyl-N,O-dimethylhydroxylamine
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Boiling Pt: 94 °C Density: 1.01 (20 °C) Flash Pt: 49 °C |
MDL Number:
MFCD00060098 CAS Number: 78191-00-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...